Learn More
Wilmad Labglass Coaxial Small Volume NMR Insert for 5mm NMR
Supplier:  Wilmad Labglass WGS5BLSP
Encompass_Preferred
The most versatile and reliable coaxial system available for various NMR experiments. General applications include small volume NMR, external referencing or locking and magnetic susceptibility determination. Ideal for variable temperature studies, all parts are made from Pyrex glass. Note: Outer tube must be ordered separately based on magnetic field strength. Note: This product is only compatible to be used with Wilmad-LabGlass 7 inch Precision NMR Sample tubes 506-PP to 542-PP
Catalog No. NC0918010
Add to cart
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'