Learn More
Wilmad Labglass Coaxial Insert for 5 mm NMR Precision Sample Tube Only
Supplier: Wilmad Labglass WGS5BL
The most versatile and reliable coaxial system available for various NMR experiments. General applications include small volume NMR, external referencing or locking and magnetic susceptibility determination. Ideal for variable temperature studies, all parts are made from Pyrex glass.