Learn More
Diapharma Group, Inc. COATEST SP FACTOR VIII
Supplier: Diapharma Group, Inc. K824086
Chromogenix Coatest® SP Factor VIII is a chromogenic assay kit for the in vitro diagnostic photometric determination of factor VIII activity in citrated plasma, such as when identifying factor VIII deficiency or monitoring patients on replacement therapy, as well as for potency estimation of FVIII concentrates. This factor VIII test kit has two measuring ranges plus validated and documented protocols for a wide range of instruments.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.