Learn More
Cole-Parmer Extech CO210 Desktop Indoor Air Quality COMonitor with Memory
Supplier: Cole-Parmer 8197371
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Monitor air quality in schools, office buildings, greenhouses, factories, and anywhere that carbon dioxide is a concern. Measures and displays carbon dioxide concentration, air temperature and humidity. Captures min/max; press RESET to clear memory. Also displays the year, month, date, and time of day. Audible alarm sounds when carbon dioxide levels exceed the set point. Activates an alarm relay that can be used for connection to an external indicating or controlling device.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.