Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Yellow, 10-425, 0.040" PTFE/Silicone/PTFE Septa
Supplier: Chemglass Life Sciences CV5075Y010
Preassembled Closure and Septa, Yellow, 10-425, 0.040" PTFE/Silicone/PTFE Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.