Learn More
Essendant Concentrated Germicidal Bleach, Regular, 121oz Bottle, 3/Carton
Supplier: Essendant CLO30966CT
Clorox Germicidal Bleach, with a concentration level of 8.25%, cleans, disinfects and deodorizes a variety of surfaces. It is EPA-registered to kill 39 organisms, including multiple strains of MRSA, Norovirus and 3 strains of influenza. Clorox Germicidal Bleach kills Clostridium difficile spores in 5 minutes on hard, nonporous surfaces when used as directed. Suitable for food-contact surfaces, Clorox Germicidal Bleach kills salmonella, E. coli and listeria. With the higher concentration level, dosage requirements for general disinfection are cut in half, providing better value and a smaller, easier to handle bottle. From Clorox Commercial Solutions, germicidal bleach is ideal for use in busy healthcare environments, offices, schools and other commercial facilities.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.