Learn More
Morwell MD Biosciences CLLGN TII CHKN SOL 10MG 2MG/ML
SDP
Supplier:  Morwell MD Biosciences 804002SOL
Encompass
Highest quality (>99%) Collagen type II (CII) protein, purified from chicken sternum, for the induction of arthritis in the Collagen-Induced Arthritis (CIA) model. Soluble 10 mg (2 mg/mL).
Catalog No. NC2467677