Learn More
United Scientific™ Clips For Jointed Glassware, Polypropylene
Prevent breakage in the lab by safely clipping jointed glassware together.
$10.30 - $15.88
Catalog Number | Mfr. No. | Color | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Description | Price | Quantity & Availability | ||||
S15805
|
United Scientific™
CLJP2932PK10 |
Red |
Pack of 10 for $15.88
|
|
|||||
S16040
|
United Scientific™
CLJP1423PK10 |
Yellow |
Pack of 10 for $10.30
|
|
|||||
S15847
|
United Scientific™
CLJP1926PK10 |
Blue |
Pack of 10 for $12.44
|
|
|||||
Description
- Plastic conical joint clips are designed to securely holdjoint size glassware together. These are non-corrosive,and will not scratch glassware.
- Manufactured from polypropylene
- Outstanding chemical resistance
- Maximum operating temperature of 135°C
- Autoclavable
Specifications
10/Pk. | |
Joint Clamp |
Warning: Cancer and reproductive harm