Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Horizontal Clip-on Badge Holder
Description
- Clear vinyl
- Built-in orientation for attachment
- Keep your identification or access badge safe and secure
- One each
Specifications
For Use With (Application) | Access cards, security badges and IDs |
Color | Clear |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction