Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cliniqa™ Liquid QC™ Cardiac Marker Control Complete
Assayed quality control material for cardiac markers
$437.00 - $438.00
Specifications
Certifications/Compliance | CE |
---|---|
Content And Storage | 2°C to 8°C |
Type | Cardiac Marker Control |
Intended Use | Assayed quality control material for cardiac markers |
Form | Liquid |
Alert:
Human source material. Treat as potentially infectious
Catalog Number | Mfr. No. | Level | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Quantity | Price | Quantity & Availability | ||||
22-100-100
|
Cliniqa™
91481 |
1 | 6 x 3 mL |
Pack of 6 for $437.00
|
|
||||
22-100-101
|
Cliniqa™
91482 |
2 | 6 x 3 mL |
Pack of 6 for $437.00
|
|
||||
22-100-102
|
Cliniqa™
91483 |
3 | 6 x 3 mL |
Pack of 6 for $438.00
|
|
||||
22-100-103
|
Cliniqa™
91484 |
1, 2, 3 | 2 x 3 x 3 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Liquid control prepared from human plasma, human serum and human proteins
- Assists in monitoring of analytical systems within clinical range
- Assayed quality control material for CK-MB, d-Dimer, hsCRP, Myoglobin, Troponin I, Troponin T and NT-proBNP
- Needs no reconstitution or frozen storage
Certifications
CE marked
Specifications
CE | |
Cardiac Marker Control | |
Liquid |
2°C to 8°C | |
Assayed quality control material for cardiac markers |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title