Learn More
Cliawaived Inc CLIAwaived, Inc. hGB Test System
Supplier: Cliawaived Inc CLIAHGB101
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
(1 Meter)The CLIAwaived, Inc Hemoglobin (Hb) Testing System is for the quantitative determination of hemoglobin in non-anticoagulated capillary whole blood or anticoagulated venous whole blood in EDTA (K2, K3, Na2) or sodium heparin. The testing system is designed for point-of-care use in primary care settings. Estimation of hematocrit is only for hemoglobin values from 12.3 to 17.5 g/dL (123 to 175 g/L).This meter is FDA Cleared and CLIA waived and for medical office use only. The CLIAwaived Hgb meter requires use of the CLIAwaived Hgb test strips. All test strips are sold separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.