Learn More
Molecular Dimensions CLEAR VUE SHEETS
Supplier: Molecular Dimensions MD601S
Protein Crystallography. ClearVue Sheets (UV friendly) 100 optically clear sealing sheets for 96-well crystallization plates. 135mm x 77mm. Full adhesive area with two perforated tabs Two end tabs for accurate positioning. Packaged in zippered bags.