Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Clear Vinyl Apron, X-Small

Simple clear vinyl apron that is perfect to protect clothes from splashes during science lab activities, washing dishes, crafts or other similar activities. Easily slips over head and ties around the back. Extra small size is suitable for children.
Supplier: Eisco™ VNAPXS
This item is not returnable.
View return policy
Description
- Simple clear vinyl apron for labs, dishwashing, or crafts
- Protects clothes from water and chemical splashes
- Fits around neck, ties in back
- Size: extra small (15 in. width, 22 in. length)
- Suitable for children
Specifications
X-Small | |
Clear | |
Children's Apron |
Vinyl | |
21 x 15 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title