Learn More
Ted Pella Inc Clear Nail Polish, sectioning and sample preparation for electron microscopy and light microscopy
Supplier: Ted Pella Inc 1147
Clear Nail Polish can be used to seal troughs and boats or tape boats for sectioning with a clean, clear edge. Clear Nail Polish can be utilized for securing eyelashes or fine hair to a wooden stick for section manipulation. The clear coating provides a universal sealant of coverslips to microscope slides for permanent storage.