Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Clear Molded Glass Type I Borosilicate Serum Vials
The utilization of type I molded glass enhances mechanical resilience, thereby minimizing the potential for particle contamination during crucial fill/finish and lyophilization stages. Vials are depyrogenated and sterilized. Crimp cap not included.
Supplier: Qorpak™ 327204
Description
- 100% inspected in clean room environment
- Vials are washed with water for injection and dried in an ISO8 environment
- Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
- Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
- Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
- Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal
Specifications
Clear | |
Borosilicate Glass | |
Sterile | |
100 mL | |
No | |
18/Pk. | |
20 mm | |
Autoclavable | |
94.5 mm | |
Serum Vial | |
Sterile |
51.6 mm | |
No | |
Single-use | |
Lyophilization | |
108/Cs. | |
Tray Packed Neck Up | |
No | |
Crimp Cap | |
108 Vials | |
No | |
Ready to Use/Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction