Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Molded Glass Type I Borosilicate Serum Vials

The utilization of type I molded glass enhances mechanical resilience, thereby minimizing the potential for particle contamination during crucial fill/finish and lyophilization stages. Vials are depyrogenated and sterilized. Crimp cap not included.

Supplier:  Qorpak™ 327206

 View more versions of this product

Catalog No. 01-672-067


Add to Cart

Description

Description

  • 100% inspected in clean room environment
  • Vials are washed with water for injection and dried in an ISO8 environment
  • Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
  • Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
  • Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
  • Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal
Specifications

Specifications

Clear
Borosilicate Glass
Sterile
10 mL
No
80/Pk.
20 mm
Autoclavable
53.5 mm
Serum Vial
Sterile
25.4 mm
No
Single-use
Lyophilization
640/Cs.
Tray Packed Neck Up
No
Crimp Cap
640 Vials
No
Ready to Use/Sterile
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit