Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clear Full Sinus Model

Full-size clear face used to display solid renditions of the paranasal and mastoid sinus cavities.
Supplier: Galloway Plastics Inc 2851
Includes: Educational card.
Description
Made of durable, high-quality materials, model provides views of sphenoid, frontal, ethmoid and maxillary sinuses.
- Model dimensions: 7 × 5.5 × 5 in. (17.8 × 14 × 12.7cm)
- Card dimensions: 8.25 × 6.25 in. (20.9 × 15.8cm)
Specifications
Used to display solid renditions of the paranasal and mastoid sinus cavities | |
Galloway Plastic | |
Anatomical Model, Clear Full Sinus |
Life size model of clear face for orientation focusing on the sinus cavities, sphenoid, frontal, ethmoid, maxillary and mastoid, educational card | |
Full-size |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title