Learn More
YSI 3P LifeScience CLEANING SOLUTION FOR GALVANIC
Supplier: YSI 3P LifeScience 205204
Cleaning solution for galvanic oxygen sensors StirrOx G, CellOx 325, DurOx 325, TA 197 Oxi, ConOx and MPP 350 Ox module, 1 bottle with 30 ml.