Learn More
Trilink Bio Technologies Inc CleanCap EGFP mRNA, 1 mg
Supplier: Trilink Bio Technologies Inc L76011000
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.