Learn More
Trilink Bio Technologies Inc CleanCap EGFP mRNA, 100 ug
Supplier:  Trilink Bio Technologies Inc L7601100
Encompass
The EGFP mRNA will express an enhanced version of the green fluorescent protein, originally isolated from the jellyfish, Aequorea victoria. EGFP is a commonly used direct detection reporter in mammalian cell culture, yielding bright green fluorescence with an emission peak at 509 nm.
Catalog No. NC1389985