Learn More
Eisco™ Class B Glass Burette with Pinch Clip
Eisco™ Class B Glass Burette with Pinch Clip designed with a single bore, detachable rubber tube, and pinch clip.
$2.70 - $9.50
Catalog Number | Mfr. No. | Capacity (Metric) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Type | Price | Quantity & Availability | ||||
S29390
![]() |
Eisco™
CH0234A |
10 mL | Pinch Clip |
Each for $6.55
|
|
||||
S29391
![]() |
Eisco™
CH0234B |
25 mL | Pinch Clip |
Each for $9.50
|
|
||||
S29392
![]() |
Eisco™
CH0234C |
50 mL | Pinch Clip |
Each for $8.65
|
|
||||
S29393
![]() |
Eisco™
CH0234D |
100 mL | Pinch Clip |
Each for $8.95
|
|
||||
S27029
![]() |
Eisco™
CH0234TIP |
Replacement Burette Clip |
Each for $2.70
|
|
|||||
Description
- Made from high quality 3.3 Borosilicate Glass
- Graduated
- Class B, DIN ISO 385 Compliant
- Single bore with detachable rubber tubing and pinch clip
Warranty and Services
1 year limited manufacturer warranty
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.