Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Supplies Class 3R Laser Pointers
Ideal for optical experiments
$10.50 - $115.00
Specifications
Product Type | Laser Pointer, Class 3R |
---|---|
For Use With (Application) | Optical Experiments |
Length (English) | 6 in. |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
S27840
|
United Scientific Supplies
RLP006 |
Red |
Each for $10.50
|
|
|||||
S27811
|
United Scientific Supplies
GLP006 |
Green |
Each for $44.50
|
|
|||||
S27797
|
United Scientific Supplies
DLP006 |
Red/Green |
Each for $115.00
|
|
|||||
Description
- Class 3R Laser Pointer with push-button switch
- Ideal for optical experiments
- Housed in rubberized aluminum
- Pen-style case with pocket clip
- Dimensions (L x D): 6 x 0.5 in.
- Maximum Output: 5 mW
- Wavelength: 532 nm
- Operates on (2) AAA batteries
- Available in Red, Green, Red/Green, and Violet
Specifications
Laser Pointer, Class 3R | |
Optical Experiments | |
6 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title