Learn More
Fisherbrand™ Class 100 Vinyl Clean Room Chair - Desk Height with Medium Back
SDPSCI_ED
Fisherbrand™ Class 100 Vinyl Clean Room Chairs with Medium Back are height adjustable for 31 in. work surfaces and feature ergonomic adjustments, clean room components, arms and self-braking casters. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
$528.00
Specifications
Arm Style 6-way adjustable up and down, in and out, twist toward or away from the user
Certifications Class 100/ISO Class 5
Foot Type Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Footrest No
Material Vinyl
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100085
SDPSCI_ED
View Documents
Taupe
Each for $528.00
Add to Cart
S0100105
SDPSCI_ED
View Documents
Marine Blue
Each for $528.00
Add to Cart
S0100110
SDPSCI_ED
View Documents
Sterling
Each for $528.00
Add to Cart
S0100111
SDPSCI_ED
View Documents
Carbon
Each for $528.00
Add to Cart
S0100112
SDPSCI_ED
View Documents
Storm
Each for $528.00
Add to Cart
S0100113
SDPSCI_ED
View Documents
Borscht
Each for $528.00
Add to Cart
Description
Description
  • Medium Back
  • Adjustable from 19 to 24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
  • Class 100/ISO Class 5 Clean Room
Specifications
Specifications
6-way adjustable up and down, in and out, twist toward or away from the user
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Vinyl
42 lb.
Yes
No
Cleanroom Seating
Class 100/ISO Class 5
No
Desk Height
Yes
Yes
Polished Cast Aluminum