Learn More
Cole-Parmer Extech EX830 1000A AC/DC True RMS Clamp/DMM/IR Thermometer
Supplier:  Cole-Parmer 2683424
Encompass_Preferred
Theses clamp-on multimeters priovide a built-in IR thermometer with laser pointer that makes troubleshooting heat sensitive areas simple. Features include data hold, Min/Max, autoranging with manual range button, auto power off, and an easy-to-read 4000 count backlit dual LCD display. Peak hold captures inrush currents and transients, and models 26834-22, -24 include true RMS and a type K thermometer probe. Units ar CAT III, 600V rated.
Model 26834-26 is CAT IV 600V rated.
Catalog No. 50-992-982
Explore available promotions