Learn More
Hach Company Clamp, Buret, Double
Supplier: Hach Company 32800
Holds two burets from micro-size to 100 mL separately without slippage or obscured graduation lines and numbers. Built-in meniscus reader. Overall size, 24 x 13 cm (9.5 x 5.2") (W x L).