Learn More
Hach Company Clamp, Buret
Supplier:  Hach Company 2087801
Encompass_Preferred
Linear polyethylene. Non-corrosive clamp holds buret firmly in place without obscuring graduations. Center clamp tightens to a standard support rod. Includes 1.25 cm (0.5") and 0.95 cm (0.4") bushings.
Catalog No. 50-199-5288