Learn More
Chemglass Life Sciences Buret Clamp
Supplier: Chemglass Life Sciences CG9002102
Buret Clamp. Clamp will hold two burets from micro to 100mL. Spring loaded mechanism adjusts simply by squeezing, yet hold buret firmly in place. Mounts directly to standard support rod with built in hook connector. Constructed of epoxy coated aluminum with vinyl jaws.