Learn More
Perkin Elmer US LLC Demountable Cell Window - NaCl, 4mm, Qty. 2
Supplier: Perkin Elmer US LLC L1271102
Circular Demountable Windows are available in a variety of window materials. Samples are placed on one window and the second window is pressed lightly on top of the first. Spacers of various thicknesses can be used to vary the cell path length. These windows require, but do not include, the Universal Demountable Cell (L1270986).