Learn More
Ted Pella Inc CIRCUIT SCRIBE CONDUCTIVE INK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ted Pella Inc 16000
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Circuit Scribe silver conductive ink pen can be used on any surface a rollerball pen can write on. Typical resistance of 1ohm/cm. Dries approximately within a minute or two. Can draw 60-200 meters depending on writing surface and speed. Non-toxic.
This item is not returnable.
View return policy