Learn More
Diapharma Group, Inc. Chromogenix Coatest SP4 Factor VIII, 200 tests
Supplier: Diapharma Group, Inc. K824094
Chromogenix Coatest SP4 Factor VIII is a chromogenic assay kit for the in vitro diagnostic photometric determination of factor VIII activity in citrated plasma.