Learn More
Thermo Scientific Thermostat Replacement Kit [1]
Supplier: Thermo Scientific 7100.0108
The CHROMELEON SINGLE EDT SE detector uses an electron capture detector to detect electronegative compounds in gas chromatography by ionizing carrier gas with a radioactive source and measuring changes in current when sample components capture electrons.
This item is not returnable.
View return policy