Learn More
MilliporeSigma™ Supelco™ ChromegaChiral™ Chiral CC2 HPLC Column, 10 μm Particle Size
ChromegaChiral™ CC2 is a modified cellulose including 3-chloro-4 methylphenyl-carbamate bonding groups coated on high purity, high performance spherical silica particles.
Supplier: MilliporeSigma™ Supelco™ ES5662
Description
This combination of bonded groups stabilizes the solubility of coated phase making for a durable phase similar to other widely used coated phases.Specifications
3 mm | |
10 μm | |
3-chloro-4 methylphenylcarbamate | |
3-chloro-4 methylphenylcarbamate Phase | |
Chiral HPLC |
10 cm | |
1000 Å | |
ChromegaChiral | |
Chiral |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
ChromegaChiral is a trademark of ES Industries