Learn More
MilliporeSigma™ Supelco™ ChromegaChiral™ Chiral CC2 HPLC Column, 10 μm Particle Size
ChromegaChiral™ CC2 is a modified cellulose including 3-chloro-4 methylphenyl-carbamate bonding groups coated on high purity, high performance spherical silica particles.
Supplier:  MilliporeSigma™ Supelco™ ES5670
Request Bulk Order
 View more versions of this product
Catalog No. 11-100-5553
Add to Cart
Description
Description
This combination of bonded groups stabilizes the solubility of coated phase making for a durable phase similar to other widely used coated phases.
Specifications
Specifications
20 mm
10 μm
3-chloro-4 methylphenylcarbamate
3-chloro-4 methylphenylcarbamate Phase
Chiral HPLC
25 cm
1000 Å
ChromegaChiral
Chiral
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
ChromegaChiral is a trademark of ES Industries