Learn More
Chondroitin sulfate, 90+%, Thermo Scientific Chemicals
Form: Powder
$27.65 - $219.65
Specifications
Chemical Name or Material | Chondroitin sulfate |
---|---|
Color | White |
MDL Number | MFCD00146419 |
Merck Index | 14,2214 |
Synonym | chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ6034103
|
Thermo Scientific Chemicals
J6034103 |
1 g |
Each
for $27.65
Save 18% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6034106
|
Thermo Scientific Chemicals
J6034106 |
5 g |
Each for $70.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6034114
|
Thermo Scientific Chemicals
J6034114 |
25 g |
Each
for $219.65
Save 23% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Chondroitin sulfate is used in a wide variety of areas of research such as bioengineering. Chondroitin sulfate may be used as a biomaterial copolymer or surface derivatization reagent in the development of drug delivery vehicles, tissue engineering devices and bioscaffolds. Chondroitin sulfate may be used for the development of biocompatible structures such as hydrogels, sponges, biofilms, microspheres and micelles. Used in the treatment of coronary atherosclerosis, myocardial ischemia, myocardial infraction. Chondroitin sulfate is a sulfated (in different positions) glycosaminoglycan (GAG), composed of repeating and alternating disaccharide units of D-glucuronic acid and N-acetylgalactosamine. Depending on the position of sulfate, Chondroitin sulfate is named as chondroitin sulfate A (sulfate on carbon 4 of the N-acetylgalactosamine (GalNAc) sugar, C (carbon 6 of the GalNAc sugar), D (carbon 2 of the glucuronic acid and 6 of the GalNAc sugar) and E (carbons 4 and 6 of the GalNAc sugar). The CAS # given is the representation of core component of this product.
SolubilitySoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications
Chondroitin sulfate | |
MFCD00146419 | |
chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid | |
KXKPYJOVDUMHGS-OSRGNVMNSA-N | |
(2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
≥90% |
White | |
14,2214 | |
Soluble in water. | |
CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O | |
24766 | |
Powder |
Safety and Handling
EINECSNumber : 232-696-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.