Learn More
Enzo Life Sciences Cholera toxin (Vibrio cholerae), (azide free) (1 mg) CAS: 9012-63-9
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Enzo Life Sciences BMLG1170001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
G protein activator. Formulation: Lyophilized from 0.05M Tris, pH 7.5, containing 200 mM NaCl, and 1 mM Na2 EDTA. Source: From Vibrio cholerae. UniProt: P01555 (A subunit), P01556 (B subunit). Handling: Handle gently and do not vortex. Store at +4°C prior to and following reconstitution. Do not freeze. Long Term Storage: +4°C