Learn More
Choice™ Polypropylene (PP) Syringe Filters
Use Choice Polypropylene Syringe Filters as an economic solution to filtration of biological samples, and for filtration of aggressive organic solutions.
$97.50 - $112.30
Specifications
Disposable | Single-use |
---|---|
Material | Polypropylene |
Material (Membrane) | Polypropylene |
Sterility | Non-sterile |
For Use With (Application) | HPLC, SPE |
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) | Prefilter | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Diameter (Metric) | Prefilter | Price | Quantity & Availability | |||
03-253-904
|
Thermo Scientific
CH2225PP |
0.2 μm | 25 mm | No |
Pack of 100 for $112.30
|
|
|||
03253916
|
Thermo Scientific
CH4525PP |
0.45 μm | 25 mm | No |
Pack of 100 for $112.30
|
|
|||
03-253-898
|
Thermo Scientific
CH2213PP |
0.2 μm | 13 mm | No |
Pack of 100 for $97.50
|
|
|||
03-253-910
|
Thermo Scientific
CH4513PP |
0.45 μm | 13 mm | No |
Pack of 100 for $97.50
|
|
|||
Description
Thermo Scientific™ Choice™ Polypropylene Syringe Filters are a good choice for filtration of biological samples, and for filtration of aggressive organic solutions. They provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.
Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect instruments and prolong column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.
Specifications- Membrane: Hydrophilic polypropylene
- Maximum operating temperature: 110 °C
- Housing: Medical grade, virgin polypropylene
- Protein- or peptide-based assays
- General HPLC analysis
- Hexane, toluene, benzene, limited resistance to dichloromethane and chloroform
Specifications
Single-use | |
Polypropylene | |
HPLC, SPE | |
100 | |
90 psi | |
Syringe Filter |
Polypropylene | |
Non-sterile | |
Solvent Resistent, Low-Extractable Polypropylene | |
No | |
Choice |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.