Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroquine Phosphate, EP, 98.5-101%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
50-63-5
C18H32ClN3O8P2
515.86
MFCD00069852
QKICWELGRMTQCR-UHFFFAOYNA-N
7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
50-63-5
C18H32ClN3O8P2
515.86
MFCD00069852
QKICWELGRMTQCR-UHFFFAOYNA-N
7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid)
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

C1960, 50-63-5, C18H26ClN3•2H3PO4

Spectrum™ Chemical Chloroquine Phosphate, EP is used as a bacteriacide, an algaecide and as an antimalarial. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Quantity 2.5 kg
Percent Purity 98.5 to 101%
Grade EP
Loss on Drying 2%
Packaging Poly Pail
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit