Learn More
Sigma Aldrich Fine Chemicals Biosciences Chloroform-d, 865-49-6, MFCD00000827, 10X0.75 mL
Supplier: Sigma Aldrich Fine Chemicals Biosciences 49427510X0.75ML
Linear Formula: CDCl3, Molecular Weight: 120.38, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS, Synonym: Deuterochloroform. Chloroform-d may be used in the synthesis of dichlorofluoromethane-d.