Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene, 1% ethanol, >99.9% (excluding ethanol), Honeywell Burdick & Jackson

Catalog Number 6001747
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
67-66-3, 64-17-5, 513-35-9
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
67-66-3, 64-17-5, 513-35-9
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
Absorbance 0.3 AU max. at 250 nm (UV)
CAS Max % 98.9900%, 1.0000%, <0.0100%
UN Number UN1888
Beilstein 1731042
Solubility Information 81.5% in Water at 20°C
Formula Weight 119.37
Percent Purity >99.9%
Grade HPLC
Density 1.49 g/cm3
Boiling Point 62°C
Quantity 6 x 1 L
Packaging Glass Bottle
Water 0.02% max.
Chemical Name or Material Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene and 1% ethanol, >99.9% (excluding ethanol)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit