Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorobenzene, CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Melting Point | -45°C (literature) |
Absorbance | max. 1.0 at 288 nm, max. 0.01 at 400 nm, max. 0.01 at 360 nm, max. 0.04 at 325 nm, max. 0.05 at 300 nm, max. 0.40 at 290 nm |
CAS Max % | 100.0000% |
Density | 1.106 g/mL at 25 °C(lit.) |
Boiling Point | 130°C to 132°C |
Quantity | 1 L |
Flash Point | 28°C |
Packaging | Glass Bottle |
Linear Formula | C6H5Cl |
UN Number | UN1134 |
Show More |
Safety and Handling
ShelfLife | 1260 days from date of manufacture |
DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Chlorobenzene |
EINECSNumber | 203-628-5 |
Recommended Storage | Room Temp |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction