Learn More
Sigma Aldrich Fine Chemicals Biosciences Cholera Toxin from Vibrio cholerae, 9012-63-9, MFCD00130776, 0.5 mg
Supplier: Sigma Aldrich Fine Chemicals Biosciences C8052.5MG
Synonym: CTX, Cholera enterotoxin, Cholergen, ~95% (SDS-PAGE), lyophilized powder. Cholera Toxin from Vibrio cholerae has been used as a positive control in cAMP (cyclic AMP) assay for enterotoxins. It has been added as a supplement in cell culture media of primary tumors and epithelial cells.
**Item is regulated in the country of origin so an End User Declaration form will be provided via email, upon receipt of the order. Form is required to be completed and returned by the end user. The lead time will be apprx. 8-10 weeks upon receipt of the completed form. Please ensure email address is available on account.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.