Learn More
Bioworld Chicken Serum, Sterile Filtered, 0.2 absolute, heat inactivated, 500 mL
Supplier:  Bioworld 306111832
Encompass_Preferred
Sterility tested.Mycoplasma tested upon request.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Heat inactivation is available if desired.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-254-258