Learn More
Rockland Immunochemicals CHICKEN RED BLOOD CELLS (RBC) 10% Washed Pooled Cells - R401-0050
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Rockland Immunochemicals R4010050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Chicken whole blood is washed to remove the platelet rich plasma, buffy coat layer, and leukocytes (WBC). Red blood cells are supplied as a 10 percent suspension in phosphate buffered saline (PBS). Chicken red blood cells are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Chicken red blood cells are perishable and are collected and processed upon receipt of your order.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.