Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Clear, PTFE/Red Rubber Septum
SDP
Supplier:  Chemglass Life Sciences CV35700011
Encompass_Preferred
Cap, Chem Snap, 11mm, Clear, PTFE/Red Rubber Septum. Chem Snap Caps may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.
Catalog No. 50-153-2556