Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ ChemoPlus™ Sterile Poly-Coated Full Coverage Blue Chemotherapy Gowns, Full Back

Catalog Number 17009991
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Sterile gowns protect healthcare personnel from exposure to toxic chemotherapy drugs during preparation, handling, and administration.

  • AAMI Level 3
  • Evaluated for resistance to permeation of 16 chemotherapy drugs per ASTM F739-20
  • Meet USP <800> standards for handling hazardous drugs
  • Reinforced critical zones include chest, sleeve seam, sleeve and front belt attachment points where direct contact with infectious material is most likely to occur
  • Full back style gowns that close and tie in the back
  • Hoop and loop neck, waist ties
Width (English) 33.5 in.
Length (Metric) 129.794 cm
Width (Metric) 85.09 cm
Length (English) 51.1 in.
Size 2X-Large
No. of Pockets None
Gender Unisex
For Use With (Application) Hazards associated with handling chemotherapy drugs
Product Type Chemotherapy Gown
Material Polyethylene-coated Polypropylene
Color Blue
Closure Type Waist Ties
Disposable Single-use
Sterility Sterile
Cuff Style Knit
Model CT5505TS
Sleeve Style Long Sleeve
Certifications/Compliance AAMI Level 3, ASTM F739-20, USP 800 Standards, FDA 510K
Autoclavable Not Autoclavable
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit