Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardinal Health™ ChemoPlus™ Universal Poly-Coated Protective Sleeve
Click to view available options
Sterility:
Non-sterile
Description
- Universal fit
- Latex-free
Specifications
Product Type | Controlled Environment Sleeves |
Size | Standard |
Construction | Poly-coated |
Material | Polyethylene-coated Polypropylene |
Color | Blue |
Sterility | Non-sterile |
Disposable | Single-use |
Closure Type | Elastic Ends |
Cuff Style | Knit Cuffs |
Model | CT5500T |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction