Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Chemistry Hardware Assortment, Deluxe
SDPSCI_ED

Most commonly used items in general chemistry labs. The Chemistry Hardware Assortment includes a comprehensive assortment of glassware, plasticware, porcelain, and other hardware.

Supplier:  United Scientific™ HRDKIT4

Catalog No. S23933


Add to Cart

Description

Description

Includes:

  • Beaker, Glass, 100 mL (1)
  • Beaker, Glass, 250 mL (1)
  • Beaker, Glass, 400 mL (1)
  • Beaker, Glass, 1000 mL (1)
  • Bottle, Wash, 125 mL, Plastic (1)
  • Clamp, Burette (1)
  • Clamp, Mohr's Pinchcock (1)
  • Crucible with Cover, 10 mL (1)
  • Cylinder, Graduated, Glass, 10 mL (1)
  • Cylinder, Graduated, Glass, 50 mL (1)
  • Cylinder, Graduated, Plastic (PP),100 mL (1)
  • Evaporating Dish, Porcelain, 35 mL (1)
  • Filter Paper, 9cm dia, pk/100 (1)
  • Flask, Boiling, 100 mL (1)
  • Flask, Boiling, 250 mL (1)
  • Flask, Erlenmeyer, Glass, 250 mL (1)
  • Flask, Erlenmeyer, Glass, 500 mL (1)
  • Flask, Volumetric, w/stopper,250 mL (1)
  • Laboratory Apron (1)
  • Mortar and Pestle, 90 mm (1)
  • Rubber Stopper, #1, Solid (6)
  • Rubber Stopper, #1, One-hole (1)
  • Rubber Stopper, #2, Solid (1)
  • Rubber Stopper, #2, One-hole (1)
  • Rubber Stopper, #2, Two-hole (1)
  • Rubber Stopper, #6, Solid (3)
  • Rubber Stopper, #6, One-hole (3)
  • Rubber Stopper, #6, Two-hole (3)
  • Spatula, Stainless Steel (1)
  • Test Tube, Glass, 16 x 150 mm (6)
  • Test Tube Brush (1)
  • Test Tube Clamp (1)
  • Test Tube Rack, 2-tier, Wooden (1)
  • Tongs, Crucible (1)
  • Triangle, Clay Pipe (1)
  • Tripod Stand, 6 in. (1)
  • Stirring Rod, Glass, 12 Pk. (1)
  • Support Stand, with 18 in. Rod (1)
  • Support Ring, 3 in. OD with Clamp (3)
  • Test Paper, Litmus, Blue (1)
  • Test Paper, Litmus, Red (1)
  • Test Paper, Cobalt Chloride (1)
  • Tubing, Glass, Assorted, One Pound (1)
  • Wire Gauze, 5 in. Square (1)

Warranty and Services

1 Year

Specifications

Specifications

Chemistry Hardware Assortment, Deluxe
5+
Doppling Bottles, Dropping Pipettes, Beakers, Evaporating Dish, Stirring Rods, Flasks, Test Tubes, Cylinders, Forcep, Tongs, Funnels, Test Tube Clamp, Wire Gauze, Crucible, Watch Glass, Scoop, Spautula, Rubber Stoppers
1 to 2 Lab Groups
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit