Learn More
Crescent Chemical Co Inc CHEMILUMINESCENCE REAGENT
Supplier: Crescent Chemical Co Inc SE42582.01
Chemiluminescence reagent for Horseradish Peroxidase Ready-to-use substrate solution for chemiluminescence detection of membrane bound antigens or nucleic acid sequences directly with Horseradish Peroxidase (HRP) or indirectly with HRP-conjugated antibodies or Streptavidin