Learn More
Haztech Systems, Inc HAZCAT 2.0 Industrial Chemical Identification Kit
Supplier: Haztech Systems, Inc KT2011
The KT2011 is a Qualitative Chemical Analysis Field Kit that identifies thousands of industrial chemicals, drug lab precursors, chemical warfare agent precursors, heavy metals, poisons, reactive oxidizers and reducers, etc. The enduser does not neeed to be a chemist. Typically takes one day to learn how to use the kit. Complete – everything needed for field testing
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.