Learn More
Global Cooling CHART RECORDER KIT,6",7DAYS
Supplier:  Global Cooling D6CHT
Encompass
Free standing/wall mountable chart recorder kit with 6 red pens, 60 paper charts and temperature probe (7 days) 6”
Catalog No. NC2054970